Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
DuPont™ Tychem™ SL 127 Series Coveralls
Shop All DuPont Personal Protection Products
Click to view available options
Size:
5X-Large
6X-Large
7X-Large
Description
DuPont Tychem 4000 delivers effective protection against a range of chemical environments. Tychem 4000, utilizes Saranex 23-P film laminated to Tyvek fabric to provide a rugged and durable fabric that resists abrasion. Tychem 4000 fabric provides at least 30 minutes of protection against >124 chemical challenges. Typical applications: chemical mixing, hazardous remediation, emergency medical response, paint spraying, and nuclear environments.Taped seams provide strong chemical resistance against heavy liquid splashes. A sewn seam is covered with a strip of compatible chemical-resistant material through heat sealing.Attached hood with elastic around face openingStorm flap covers zipper which can be sealed by the wearer with adhesive strip to prevent intrusion at zipperElastic opening for tighter fit at wristElastic opening for tighter fit at ankle

Specifications
Specifications
Size | 6X-Large |
Material | Tychem |
Color | White |
Ankle Style | Elastic |
Neck Style | Hooded |
Hood Type | Attached |
Closure Type | Front Zipper |
Seam Type | Taped |
Description | Attached hood |
For Use With (Application) | Waste management, hazardous response and nuclear,environments |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction