Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DuPont™ Tychem™ SL Coveralls

Catalog No. p-315884 Shop All DuPont Personal Protection Products
Click to view available options
Size:
X-Large
3X-Large
This item is not returnable. View return policy
This item is not returnable. View return policy
DuPont Tychem 4000 delivers effective protection against a range of chemical environments. Tychem 4000, utilizes Saranex 23-P film laminated to Tyvek fabric to provide a rugged and durable fabric that resists abrasion. Tychem 4000 fabric provides at least 30 minutes of protection against >124 chemical challenges. Typical applications: chemical mixing, hazardous remediation, emergency medical response, paint spraying, and nuclear environments.Taped seams provide strong chemical resistance against heavy liquid splashes. A sewn seam is covered with a strip of compatible chemical-resistant material through heat sealing.Laydown collarStorm flap covers zipper which can be sealed by the wearer with adhesive strip to prevent intrusion at zipperElastic opening for tighter fit at wristElastic opening for tighter fit at ankle
TRUSTED_SUSTAINABILITY

Specifications

Size X-Large
Material Tychem
Color White
Waist Style Waist Ties
Cuff Style Elastic
Closure Type Snap
Seam Type Taped
Description Elastic wrist, 44 in. long, neck loop with snap and waist ties
Size (English) Chest 42.75 to 46.25 in.
Size (Metric) Chest 108.58 to 117.47 cm
Sleeve Style Long sleeve
Thickness 12 mil
For Use With (Application) Chemical mixing, hazardous remediation, emergency medical response, paint spraying and nuclear environments
Length (English) 45 in.
Length (English) Sleeve 32.25 in.
Length (Metric) 2.812 cm
Length (Metric) Sleeve 81.91 cm
Height (English) 5.8 to 6.2 ft.
Height (Metric) 1.76 to 1.88 m
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.