Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DuPont™ Tychem™ 4000 Series 127 Coveralls

Catalog Number 19120694C
Click to view available options
Size:
Medium
Large
X-Large
2X-Large
3X-Large
4X-Large
5X-Large
6X-Large
7X-Large
This item is not returnable. View return policy
This item is not returnable. View return policy

DuPont™ Tychem™ 4000 Coveralls provide at least 30 minutes of protection against >124 chemical challenges in a range of chemical environments.

DuPont™ Tychem™ 4000 delivers effective protection against a range of chemical environments. Tychem™ 4000, utilizes Saranex™ 23-P film laminated to Tyvek™ fabric to provide a rugged and durable fabric that resists abrasion. Tychem™ 4000 fabric provides at least 30 minutes of protection against >124 chemical challenges. Typical applications: chemical mixing, hazardous remediation, emergency medical response, paint spraying, and nuclear environments.

  • Tightly sewn seam is covered with garment fabric to reinforce seam and reduce potential for particle penetration
  • Attached hood with elastic around face opening
  • Storm flap covers zipper which can be sealed by the wearer with adhesive strip to prevent intrusion at zipper
  • Elastic opening for tighter fit at wrist
  • Elastic opening for tighter fit at ankle

Compliance

Meets ASTM™ D3776, ASTM D5034, ASTM D1777, ASTM F739, ASTM D3786; 16 CFR 1610 Standard for Flammability of Clothing Textiles; MIL-STD-282 requirements.

TRUSTED_SUSTAINABILITY
Size 3X-Large
Material Tychem
Color White
Ankle Style Elastic
Cuff Style Elastic
Neck Style Hooded
Closure Type Zipper
Seam Type Bound
Certifications/Compliance ASTM D3776, ASTM D5034, ASTM D1777, ASTM F739, ASTM D3786, 16 CFR 1610, MIL-STD-282
Size (English) Chest 47.25 to 50.75 in.
Size (English) Inseam 31 in.
Size (Metric) Chest 120 to 128.9 cm
Thickness 12 mil
For Use With (Application) Chemical mixing, hazardous remediation, emergency medical response, paint spraying and nuclear environments
Length (English) Sleeve 38.5 in.
Length (Metric) Sleeve 97.79 cm
Width (English) Chest 30.25 in.
Height (English) 6.2 to 6.4 ft.
Height (Metric) 1.88 to 1.95 m
Weight (English) 0.22 lb.
Weight (Metric) 0.1 kg
Show More Show Less
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit