Learn More
DuPont™ Tychem™ ThermoPro Combo Suits
Shop All DuPont Personal Protection Products
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
3X-Large
4X-Large
Color:
Gray
Orange
Description
Includes
- Jacket with taped seams, mandarin collar, elastic wrists, front zipper closure and double storm flap
- Bib Overall with taped seams and adjustable webbing straps with closure
- Constructed for heavy use, yet lightweight and easy to wear
- At least 30 minutes of protection against more than 180 chemical challenges
- Arc rating of 15.22 cal/cm Ebt
Specifications
Specifications
| Color | Orange |
| Size | Large |
| Ankle Style | Open |
| Collar Type | Mandarin |
| Cuff Style | Elastic |