Learn More
DuPont™ Tychem™ ThermoPro Combo Suits
Catalog No. 19120958 Shop All DuPont Personal Protection Products
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
3X-Large
4X-Large
Color:
Gray
Orange
This item is not returnable. View return policy
This item is not returnable. View return policy
Includes
  • Jacket with taped seams, mandarin collar, elastic wrists, front zipper closure and double storm flap
  • Bib Overall with taped seams and adjustable webbing straps with closure
Protection from chemicals, flash fire and electrical arc
  • Constructed for heavy use, yet lightweight and easy to wear
  • At least 30 minutes of protection against more than 180 chemical challenges
  • Arc rating of 15.22 cal/cm Ebt
TRUSTED_SUSTAINABILITY
Specifications
Color Gray
Size Medium
Ankle Style Open
Collar Type Mandarin
Cuff Style Elastic
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.