Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DuPont™ Tychem™ ThermoPro™ Coveralls

Catalog No. 19101354 Shop All DuPont Personal Protection Products
Click to view available options
Color:
Gray
Orange
Material:
Tychem
Size:
Small
Medium
Large
X-Large
2X-Large
3X-Large
4X-Large
5X-Large
Certifications/Compliance:
NFPA 1992, NFPA 2112, HRC 2, NFPA 70E™
NFPA 1992, NFPA 2112, HRC 2, NFPA 70E™, Berry Amendment
This item is not returnable. View return policy
Do not use for protection against continuous thermal loads, molten metal splash, hot liquids or steam. Do not wear for structural or proximity fire fighting.
This item is not returnable. View return policy
Do not use for protection against continuous thermal loads, molten metal splash, hot liquids or steam. Do not wear for structural or proximity fire fighting.

Provides chemical, flash fire and electric arc protection

Coveralls combine the chemical protection of DuPont Tychem with the flame and arc flash protection of DuPont Nomex™ into a single garment. The fabric delivers permeation protection against a broad range of toxic industrial chemicals and even chemical warfare agents.

  • Provide at least eights hours barrier to 20 of the 21 chemicals found in ASTM™ F1001
  • Attached respirator-fit hood with drawstring
  • Zipper front with Velcro™ fastener double storm flaps fastened by flame-resistant treated polyamide hook-and-loop material
  • Elastic at the wrists and ankles
  • Attached socks and boot flaps
  • Available in high-contrast orange or low-visibility gray
Specifications
  • Arc rating:15.2 cal/cm2
  • Thickness: 34mil
  • Basis Weight: 841 oz./yd.2
  • Ball Burst: 167 lbf
  • Tear Resistance–Trap Tear (MD/CD): 25/32 lbf
  • Breaking Strength–Grab (MD/CD): 155/170 lbf
  • Wearing Apparel Flammability: Class 1

† For electric arc hazard exposures, wear the proper arc rated garment system as dictated by an electric arc hazard analysis.

TRUSTED_SUSTAINABILITY

Specifications

Size Medium
Material Tychem
Color Orange
Ankle Style Attached Socks, Boot Overflaps
Cuff Style Elastic
Neck Style Hooded
Hood Type Respirator Fit (Drawstring)
Closure Type Zipper with Double Storm Flap, Hook and Loop Seal
Seam Type Taped
Arc Flash Rating 15.2 Calories/cm2
Certifications/Compliance NFPA 1992, NFPA 2112, HRC 2, NFPA 70E™, Berry Amendment
Description Respirator fit, drawstring hood, elastic wrist, attached socks with outer boot flaps, double storm flap with hook and loop closure
Size (English) Chest 34.75 to 38.25 in.
Size (English) Inseam 28 in.
Size (Metric) Chest 88.26 to 97.15 cm
Thickness 34 mil
For Use With (Application) Fire departments, industrial fire brigades, clandestine lab investigation, industrial chemical processing plants
Length (English) Sleeve 34 in.
Length (Metric) Sleeve 86.3 cm
Width (English) Chest 24 in.
Height (English) 5.3 to 5.7 ft.
Height (Metric) 1.6 to 1.73 m
Weight (English) 0.51 lb.
Weight (Metric) 0.23 kg
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.