Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
DuPont™ Tychem™ ThermoPro™ Sleeveless Orange Aprons
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
Description
- Tychem™ ThermoPro for triple hazard protection
- Sleeveless apron design provides frontal body protection
- For flash fire and electric arc hazards, partial body protective Tychem ThermoPro garments must be used in conjunction with primary flame resistant clothing that is rated for the fire/arc hazard.
- Sewn with Nomex™ thread
- Designed with Nomex waist strap and flame resistant (FR) nylon buckle
- Designed with end-user input for laboratory applications
- Ideal for use on top of an existing flame resistant (FR) garment where extra chemical protection is required
- Made in the USA
Compliance
North American Free Trade Agreement (NAFTA) and Trade Agreement Act (TAA).

Specifications
Specifications
Size | Small |
Color | Orange |
For Use With (Application) | HazMat Teams in Fire Departments, Industrial Fire Brigades, Clandestine Lab Investigation, Industrial Chemical Processing Plants, Laboratories |
Seam Type | Taped |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction