Learn More
Description
- Tychem™ ThermoPro for triple hazard protection
- Sleeveless apron design provides frontal body protection
- For flash fire and electric arc hazards, partial body protective Tychem ThermoPro garments must be used in conjunction with primary flame resistant clothing that is rated for the fire/arc hazard.
- Sewn with Nomex™ thread
- Designed with Nomex waist strap and flame resistant (FR) nylon buckle
- Designed with end-user input for laboratory applications
- Ideal for use on top of an existing flame resistant (FR) garment where extra chemical protection is required
- Made in the USA
Compliance
North American Free Trade Agreement (NAFTA) and Trade Agreement Act (TAA).
Specifications
Specifications
| Size | Large |
| Color | Orange |
| For Use With (Application) | HazMat Teams in Fire Departments, Industrial Fire Brigades, Clandestine Lab Investigation, Industrial Chemical Processing Plants, Laboratories |
| Seam Type | Taped |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.