Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DuPont™ Tychem™ ThermoPro™ TP198T Coveralls

Catalog No. 19101308
Click to view available options
Certifications/Compliance:
NFPA 1992, NFPA 2112, HRC 2, NFPA 70E™
NFPA 1992, NFPA 2112, HRC 2, NFPA 70E™, Berry Amendment
Color:
Gray
Orange
Size:
Small
Medium
Large
X-Large
2X-Large
3X-Large
4X-Large
5X-Large
Height (English):
5 to 5.7 ft.
5.3 to 5.7 ft.
5.5 to 5.9 ft.
5.8 to 6.2 ft.
6 to 6.4 ft.
6.2 to 6.4 ft.
6.4 to 6.7 ft.
6.7 to 6.10 ft.
This item is not returnable. View return policy
This item is not returnable. View return policy

ThermoPro coveralls offer superior protection against fire, electricity, and toxic chemicals.

ThermoPro coveralls are secure in fit to seal out contaminants

  • Respirator fit hood with drawstrings
  • Front zipper with storm flap, hook and loop secure
  • Elastic wrists
  • Open, hemmed ankles
  • Provides at least one hour breakthrough protection against 20 of the 21 standard ASTM F1001 chemicals
  • At least 30 minutes of protection against 182 chemical challenges

  • Protects against flash fire, electric arc and liquid-chemical splash, wide range of toxic and chemical warfare agents
  • HazMat teams in fire departments
  • Industrial fire brigades
  • Clandestine lab investigation
  • Industrial chemical processing plants

Certifications

  • NFPA 1992 (2012)
  • NFPA 2112 (2012)
  • Exceeds NFPA 70E Hazard Risk Category 2

TRUSTED_SUSTAINABILITY

Specifications

Size Large
Material HDPE
Color Gray
Ankle Style Open
Cuff Style Elastic
Neck Style Hooded
Closure Type Hook and Loop
Seam Type Taped
Certifications/Compliance NFPA 1992, NFPA 2112, HRC 2, NFPA 70E™, Berry Amendment
Description Fit hood, elastic wrist, open ankle, with respirator; HDPE Tychem™ ThermoPro construction
Size (English) Chest 38.75 to 42.25 in.
Size (English) Inseam 27.5 in.
Size (Metric) Chest 98.42 to 107.31 cm
For Use With (Application) Fire departments, industrial fire brigades, clandestine lab investigation, industrial chemical processing plants
Length (English) Sleeve 35 in.
Length (Metric) Sleeve 88.9 cm
Width (English) Chest 26 in.
Height (English) 5.5 to 5.9 ft.
Height (Metric) 1.67 to 1.8 m
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.