Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DuPont™ Tyvek™ IsoClean™ Clean Processed Frocks
GREENER_CHOICE

Catalog No. 17990525
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
3X-Large
4X-Large
Length (English):
41 in.
42.75 in.
45 in.
47 in.
53.75 in.
55 in.
57 in.
Size (English) Chest:
32 to 36 in.
36 to 38 in.
38 to 40 in.
40 in to 43 in.
43 to 46 in.
46 to 52 in.
This item is not returnable. View return policy
Should not be worn around open flame, heat source, sparks or ignition sources.
This item is not returnable. View return policy
Should not be worn around open flame, heat source, sparks or ignition sources.

Frock provides an inherent barrier to particles, microorganisms and non-hazardous light liquid splash. Clean processed garments offer lowest level of particle shedding within DuPont product portfolio.

  • Serged seams have multiple interlocking threads that are sewn around the raw edges of garment material to create a strong, stress-resistant seam
  • Elastic openings for tighter fit at wrist
  • Bound neck for lower particle shedding
  • Front zipper closure for easy donning and doffing
  • Raglan sleeve design for greater range of motion and comfort
  • A-line silhouette

 

TRUSTED_SUSTAINABILITY

Specifications

Product Type Controlled Environments Frock
Size 3X-Large
Material Flashspun Polyethylene
Color White
Sterility Non-sterile
Closure Type Zipper
Cuff Style Elastic
Seam Type Serged
Collar Type Bound
Size (English) Chest 43 to 46 in.
Size (Metric) Chest 109.2 to 116.8 cm
Sleeve Style Raglan Sleeve
Packaging Type Individually Packaged
Length (English) 55 in.
Length (English) Sleeve 44.25 in.
Show More Show Less

Adhere to
Safety Regulations and Best Practices

Take advantage of a range of Safety Services from our trusted suppliers to help ensure the safety and compliance of your people, processes, products, and facilities. Services include assessments, training, equipment maintenance, safety programs, and more.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.