Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DuPont™ Tyvek™ IsoClean™ Serged Seam Cleanroom Frocks
GREENER_CHOICE

Catalog No. 17988069 Shop All DuPont Personal Protection Products
Click to view available options
Size:
Medium
Large
X-Large
2X-Large
3X-Large
4X-Large
Sterility:
Non-sterile
Sterile
Packaging Type:
Bulk Packaged
Individually Packaged
This item is not returnable. View return policy
Garments made of Tyvek™ should not be used around heat, flames, sparks or in potentially flammable or explosive environments.
This item is not returnable. View return policy
Garments made of Tyvek™ should not be used around heat, flames, sparks or in potentially flammable or explosive environments.

Available in sterile or non-sterile versions, Tyvek™ cleanroom frocks provide an inherent barrier to particles, microorganisms, and non-hazardous light liquid splash in ISO 5 (Class 100) and above environments.

  • Serged seams have multiple interlocking threads that are sewn around the raw edges of garment material to create a strong, stress-resistant seam.
  • Elastic opening for tighter fit at wrist
  • Mandarin collar with snap for secure fit
  • Body-length jacket design with front snap closures for easy donning and doffing
  • Raglan sleeve design for greater range of motion and comfort.
  • Generous sizing
  • Non-sterile versions are bulk packaged in double transparent poly liners
  • Sterile versions are individually packaged
  • Manufactured under specifications that do not contain natural rubber latex
  • Sterile frocks are gamma sterilized to SAL of 10-6
TRUSTED_SUSTAINABILITY

Specifications

Product Type Controlled Environments Frock
Size Large
Material Polyethylene
Color White
Disposable Single-use
Sterility Non-sterile
Closure Type Snaps
Cuff Style Elastic
Seam Type Serged
Collar Type High Mandarin
Size (English) Chest 36 to 38 in.
Sleeve Style Raglan Sleeve
Packaging Type Bulk Packaged
Length (English) 45.75 in.
Length (Metric) 116.2 cm
Length (English) Sleeve 38.25 in.
Length (Metric) Sleeve 97.15 cm
Cleanroom Class Class 100 (ISO 5)
Show More Show Less

Adhere to
Safety Regulations and Best Practices

Take advantage of a range of Safety Services from our trusted suppliers to help ensure the safety and compliance of your people, processes, products, and facilities. Services include assessments, training, equipment maintenance, safety programs, and more.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.