Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DuPont™ Tyvek™ IsoClean™ Series 105 Hooded Coveralls with Attached Boots, Bulk Packaged
GREENER_CHOICE

Catalog No. 17685201 Shop All DuPont Personal Protection Products
Click to view available options
Size:
Medium
Large
X-Large
2X-Large
3X-Large
Height (English):
5.11 to 6.3 ft.
5.5 to 5.10 ft.
5.8 to 6 ft.
6.1 to 6.5 ft.
6.2 to 6.6 ft.
Height (Metric):
155.75 to 192.04 cm
167.64 to 155.44 cm
176.78 to 182.88 cm
185.92 to 198.12 cm
188.97 to 201.16 cm
This item is not returnable. View return policy
This item is not returnable. View return policy

IsoClean delivers an ideal balance of protection, durability, and comfort. Bulk packaged in double transparent poly liners.

Light and durable coveralls provide coverage, including feet, while staying comfortable. Use in controlled environments with high standards for particle and microbiological contamination control.

  • Spinning process creates barrier to particles, micro-organisms and nonhazardous liquids
  • Lightweight, comfortable and durable
  • Lowest level particle shedding
  • Recyclable
  • Serged seams
  • Standard hood and elastic opening with elastic wrists and ankles
  • Attached thumb loops
  • Zipper closure
  • Attached boots, PVC sole
  • Bulk packaged, nonsterile
  • 25 coveralls per case

Should not be worn around open flame, heat source, sparks or ignition sources.

Pharmaceutical, biotech, medical device, electronic settings

TRUSTED_SUSTAINABILITY

Specifications

Product Type Cleanroom Coveralls
Size Large
Material Flashspun Polyethylene
Color White
Disposable Single-use
Ankle Style Elastic, Attached Boots
Cuff Style Thumb Loop
Neck Style Hooded
Hood Type Elastic
Closure Type Zipper
Seam Type Serged
Sterility Non-sterile
Description Elastic wrist and ankle, standard hood with elastic opening, set sleeves, attached thumb loops, attached boot with PVC soles
Size (English) Chest 38 to 40 in.
Size (English) Inseam 29 in.
Size (Metric) Chest 96.52 to 101.6 cm
Size (Metric) Inseam 73.6 cm
Sleeve Style Set
Thickness 5.4 mil
For Use With (Application) Barrier to dry particles, microorganisms, and non-hazardous liquids in controlled environments
Length (English) Sleeve 35 in.
Length (Metric) Sleeve 88.9 cm
Width (English) Chest 25.75 in.
Height (English) 5.8 to 6 ft.
Height (Metric) 176.78 to 182.88 cm
Packaging Bulk
Cleanroom Class Class 1000 (ISO 6)
Show More Show Less

Adhere to
Safety Regulations and Best Practices

Take advantage of a range of Safety Services from our trusted suppliers to help ensure the safety and compliance of your people, processes, products, and facilities. Services include assessments, training, equipment maintenance, safety programs, and more.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.