Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
DuPont™ Tyvek™ IsoClean™ Series 226 Smocks
Click to view available options
Length (English):
36.25 in.
37.25 in.
38.25 in.
39.25 in.
Size:
Small
Medium
Large
X-Large
Description
IsoClean is excellent for controlled environments to protect against particulates and other contaminants in a laboratory setting.
- Spinning process creates barrier to particles, micro-organisms, and nonhazardous liquids
- Lightweight, comfortable and durable
- Lowest level particle shedding
- Serged seams
- Bound neck
- Three pockets
- Elastic wrists
- Wraps around front, ties at waist
Controlled environments, pharmaceutical, biotech, medical device and electronic manufacturing facilities

Specifications
Specifications
Size | Medium |
Material | Tyvek |
Color | White |
Seam Type | Serged |
Size (English) Chest | 32 to 36 in. |
Size (Metric) Chest | 81 to 91 cm |
Length (English) | 37.25 in. |
Length (Metric) | 94.61 cm |
Length (English) Sleeve | 34 in. |
Length (Metric) Sleeve | 86.36 cm |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction