Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DuPont™ Tyvek™ IsoClean™ Coverall (all)
GREENER_CHOICE

Catalog No. p-4258140 Shop All DuPont Personal Protection Products
Change view
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
3X-Large
4X-Large
5X-Large
Height (English):
5 to 5.7 ft.
5.11 to 6.3 ft.
5.5 to 5.10 ft.
5.8 to 6 ft.
6.1 to 6.5 ft.
6.2 to 6.6 ft.
6.3 to 7 ft.
6.4 to 7.2 ft.
Height (Metric):
152.4 to 173.73 cm
155.75 to 192.04 cm
167.64 to 155.44 cm
176.78 to 182.88 cm
185.92 to 198.12 cm
188.97 to 201.16 cm
192.02 to 213.3 cm
195.07 to 219.45 cm
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size Height (English) Height (Metric)
17-987-991 Small 5 to 5.7 ft. 152.4 to 173.73 cm
17-987-992 Medium 5.5 to 5.10 ft. 167.64 to 155.44 cm
17-987-993 Large 5.8 to 6 ft. 176.78 to 182.88 cm
17-987-994 X-Large 5.11 to 6.3 ft. 155.75 to 192.04 cm
17-987-995 2X-Large 6.1 to 6.5 ft. 185.92 to 198.12 cm
17-987-996 3X-Large 6.2 to 6.6 ft. 188.97 to 201.16 cm
17-987-997 4X-Large 6.3 to 7 ft. 192.02 to 213.3 cm
17-987-998 5X-Large 6.4 to 7.2 ft. 195.07 to 219.45 cm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
This item is not returnable. View return policy
Garments made of Tyvek should not be used around heat, flames, sparks or in potentially flammable or explosive environments.
Catalog No. 17-987-991 Supplier DuPont™ Supplier No. IC253BWHSM00250B
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy
Garments made of Tyvek should not be used around heat, flames, sparks or in potentially flammable or explosive environments.

Tyvek coveralls provide an inherent barrier to particles, microorganisms, and non-hazardous light liquid splash. Bound seams reduce the potential for particle penetration.

  • For use in controlled environments with high standards for particle and microbiological contamination control:
    • Pharmaceutical
    • Biotech
    • Medical Device
    • Electronics
  • Light and durable
  • Spun polyethylene creates barrier to particles, micro-organisms, and non-hazardous liquids
  • Lowest level particle shedding
  • Bound seams and neck
  • Covered elastic wrists and ankles with dolman sleeves
  • Zipper closure
  • Should not be worn around open flame, heat source, sparks or ignition sources
TRUSTED_SUSTAINABILITY

Specifications

Product Type Cleanroom Coveralls
Size Small
Gender Unisex
Material Tyvek
Color White
Disposable Single-use
Ankle Style Covered Elastic
Cuff Style Covered Elastic
Neck Style Collared
Closure Type Zipper
Seam Type Bound
Sterility Non-sterile
Collar Type Bound
Size (English) Chest 32 to 36 in.
Size (English) Inseam 29.25 in.
Size (Metric) Chest 81.28 to 91.44 cm
Size (Metric) Inseam 74.2 cm
Sleeve Style Dolman
Thickness 5.4 mil
For Use With (Application) Barrier to Dry Particles, Microorganisms, and Non-Hazardous Liquids in Controlled Environments
Length (English) Sleeve 34.25 in.
Length (Metric) Sleeve 86.9 cm
Height (English) 5 to 5.7 ft.
Height (Metric) 152.4 to 173.73 cm
Packaging Bulk
Cleanroom Class Class 1000 (ISO 6)
Show More Show Less

Adhere to
Safety Regulations and Best Practices

Take advantage of a range of Safety Services from our trusted suppliers to help ensure the safety and compliance of your people, processes, products, and facilities. Services include assessments, training, equipment maintenance, safety programs, and more.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.