Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DuPont™ Tyvek™ IsoClean™ Boot Covers
BIOPHARMA

Catalog No. p-3834077 Shop All DuPont Personal Protection Products
Change view
Click to view available options
Size:
Small
Medium
Large
X-Large
Seam Type:
Bound
Serged
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size Seam Type
17-988-092 Small Serged
17-988-093 Medium Serged
17-988-094 Large Serged
17-988-095 X-Large Serged
17-988-098 Medium Bound
17-988-099 Large Bound
19-813-220 X-Large Bound
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
This item is not returnable. View return policy
Garments made of Tyvek™ should not be used around heat, flames, sparks or in potentially flammable or explosive environments.
Catalog No. 17-988-092 Supplier DuPont™ Supplier No. IC457SWHSM01000S
Only null left
Add to Cart
Edge
Add to Cart
This item is not returnable. View return policy
Garments made of Tyvek™ should not be used around heat, flames, sparks or in potentially flammable or explosive environments.

Sterile boot covers create an inherent barrier to dry particles, microorganisms, and nonhazardous liquids.

  • Garments are gamma sterilized to SAL of 10-6.
  • Serged seams have multiple interlocking threads that are sewn around the raw edges of garment material to create a strong, stress-resistant seam.
  • Pleated rayon outer facing
  • 18 in. high boot cover with covered elastic top
  • PVC soles provides enhanced skid-resistance and durability
  • Full traceability on all sterilized apparel with Certificates of Sterility
  • D14238474
  • Garments are gamma sterilized to SAL of 10-6.
  • Serged seams have multiple interlocking threads that are sewn around the raw edges of garment material to create a strong, stress-resistant seam.
  • Pleated rayon outer facing
  • 18 in. high boot cover with covered elastic top
  • PVC soles provides enhanced skid-resistance and durability
  • Full traceability on all sterilized apparel with Certificates of Sterility
  • D15568559
TRUSTED_SUSTAINABILITY

Specifications

Size Small
Material Tyvek
Latex Free Yes
Color White
Sterility Sterile
Closure Type Elastic Top with Ankle Ties
Sole Type Anti-skid
Seam Type Serged
Thickness 5.4 mil
Length (English) 10.5 in.
Length (Metric) 26.67 cm
Height (English) 18.5 in.
Height (Metric) 46.99 cm
Show More Show Less

Adhere to
Safety Regulations and Best Practices

Take advantage of a range of Safety Services from our trusted suppliers to help ensure the safety and compliance of your people, processes, products, and facilities. Services include assessments, training, equipment maintenance, safety programs, and more.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.