Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DuPont™ Tyvek ™ Micro-Clean ™ 2-1-2 Coveralls, Sterile
BIOPHARMA

Catalog No. 17989606 Shop All DuPont Personal Protection Products
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
3X-Large
4X-Large
5X-Large
This item is not returnable. View return policy
This item is not returnable. View return policy

Ideal protection against micro-organisms and particulates in controlled environments.

Coveralls feature Micro-Clean™ with 2-1-2 polymeric resin coating that allows for thorough protection and reduction in static.

  • Made from Tyvek™ flash spun polyolefin protective material to create a barrier to particulates, nonhazardous liquids and micro-organisms
  • Available clean processed (individually packaged) or sterilized via gamma irradiation
  • Coated in 2-1-2 polymeric resin
  • Antistatic
  • Bound neck and seams with zipper closure
  • Dolman sleeves
  • Elastic wrists and ankles

TRUSTED_SUSTAINABILITY

Specifications

Product Type Cleanroom Coveralls
Size 4X-Large
Material Tyvek™
Color Blue
Ankle Style Covered Elastic
Cuff Style Covered Elastic
Neck Style Collared
Closure Type Zipper
Seam Type Bound
Sterility Sterile
Coating Polymeric Resin
Collar Type Bound
Model CC252BBU
Size (English) Chest 52 to 58 in.
Size (English) Inseam 36 in.
Size (Metric) Chest 132.08 to 147.32 cm
Size (Metric) Inseam 91.4 cm
Sleeve Style Dolman
Thickness 5 mil
For Use With (Application) Barrier to dry particles and microorganisms
Length (English) Sleeve 47.25 in.
Length (Metric) Sleeve 120 cm
Height (English) 6.3 to 7 ft.
Height (Metric) 192.02 to 213.3 cm
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.