Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DWK Life Sciences DURAN™ 5.8 mm Connector and 0.2 μm Syringe Disk Filter
SDP

Catalog No. 09841265 Shop All DWK Life Sciences Products
Change view
Click to view available options
Pore Size:
0.2 μm
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Pore Size
09-841-265 0.2 μm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 09-841-265 Supplier DWK Life Sciences Supplier No. 1129825
Add to Cart
Add to Cart

Syringe Filter (0.2 μm) with connector, non-sterile 0.2 μm filter and female Luer Slip to 5.8 mm male connector, with O-ring seal.

  • Accessory for use with DURAN™ Screw Cap GL 45 with 2-hose connector (No. 293102807)
  • Consists of: Syringe Filter (0.2 μm) with connector, non-sterile 0.2 μm filter and female Luer Slip to 5.8mm male connector, with O-ring seal
TRUSTED_SUSTAINABILITY

Specifications

Autoclavable Autoclavable
Connectors Female Luer Slip to 5.8 mm Male Connector
Filter Type Syringe Filter
For Use With (Equipment) GL 45 Screw Cap with 2-hose Connector
Material Polypropylene, EPDM
Pore Size 0.2 μm
Sterility Non-sterile
For Use With (Application) Sterile fluid transfer in and out of lab bottles; Gas transfer in and out of lab bottles; Fluid transfer bewteen lab bottles; Supply of reagents to instruments
Includes O-ring Seal
Quantity 2/Pk.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.