Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DWK Life Sciences DURAN™ GL 25 Screw Caps for Laboratory Glass Bottles
SDP

Catalog No. 09841262
Click to view available options
Color:
Blue
Red
Material:
PBT
Polypropylene

High leak tightness through use of PTFE coated silicone seal. More chemically resistant than PP screw cap.

  • Thread style: GL 25
TRUSTED_SUSTAINABILITY

Specifications

Color Red
Material PBT
Thread Size (Metric) GL 25
Product Type Screw Cap with PTFE-protected Seal
For Use With (Equipment) Laboratory glass bottles
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.