Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DWK Life Sciences DURAN™ GLS 80™ Wide Mouth Bottle Connection System, Screw Cap GLS 80™, 4-Ports, GL 18 Thread
SDP

Catalog No. 09841146 Shop All DWK Life Sciences Products
Click to view available options
Description:
Insert for screw cap GL 18, ID 10.0 mm
Insert for screw cap GL 18, ID 3.2 mm
Insert for screw cap GL 18, ID 6.0 mm
Insert for screw cap GL 18, ID 8.0 mm
This item is not returnable. View return policy
This item is not returnable. View return policy

Flexible GLS 80™ connection system for the use of semi-rigid hoses.

Flexible GLS 80™ cap connection system for use with semi-rigid tubing, suitable for DURAN™ GLS 80™ laboratory and media bottles. Five different tube diameters (3.2 mm, 6.0 mm, 8.0 mm, 10.0 mm and 12.0 mm) can be connected. Sterile pressure equalisation through membrane liter possible. Unused ports can be provided with a blind cap. Exemplary applications: Safe transfer of liquid media within a closed and sterile system.

  • The flexible modular system allows individual combination of the various components according to requirements and application
  • Five different hose diameters (3.2 mm; 6.0 mm; 8.0 mm; 10.0 mm and 12.0 mm) can be connected
  • NS 29/32 connection in the middle for tight closure
  • Sterile pressure equalisation through membrane filter possible
  • Unused ports can be fitted with a GL 18 blind cap
  • Compatible with all GLS 80 threads and therefore versatile
TRUSTED_SUSTAINABILITY

Specifications

Description Insert for screw cap GL 18, ID 10.0 mm
For Use With (Application) Fluid transfer; Supply of liquid into bioreactors; Supply of reagents to instruments; Photo-bioreactor
Material Polypropylene, PTFE
Quantity 1 Ea.
Autoclavable Autoclavable
Diameter (Metric) 87 mm
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.