Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
DWK Life Sciences DURAN™ Reaction Vessels
Shop All DWK Life Sciences Products

Click to view available options
Capacity (Metric):
250 mL
2000 mL
Description
- With groove
- For vacuum use
Specifications
Specifications
| Capacity (Metric) | 2000 mL |
| Material | Glass |
| For Use With (Application) | Vacuum use |
| Includes | Groove |
| Product Type | Flat flange reaction vessel with groove |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction