Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
DWK Life Sciences DURAN™ Tube with Screw Thread, with DIN thread

The threaded tube is used for further processing in glass components.
Supplier: DWK Life Sciences 248380209
Description
This threaded tube has a screw thread and can be incorporated into glass components by a further processing step.
- With DIN thread
Specifications
| Borosilicate Glass (3.3) | |
| 10/Pk. | |
| Autoclavable | |
| 1.8 mm |
| Shoring in glass apparatuses | |
| GL 25 | |
| 22 mm |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction