Learn More
DWK Life Sciences DURAN™ Tube with Screw Thread, with DIN thread
The threaded tube is used for further processing in glass components.
$87.00 - $295.87
Specifications
| Material | Borosilicate Glass (3.3) |
|---|---|
| For Use With | Shoring in glass apparatuses |
| Autoclavable | Autoclavable |
| Catalog Number | Mfr. No. | Thread Size | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Thread Size | Price | Quantity & Availability | |||||
09-841-138
![]() |
DWK Life Sciences
248350103 |
GL 45 |
Case for $87.00
|
|
|||||
09-841-139
![]() |
DWK Life Sciences
248380209 |
GL 25 |
Case of 10 for $161.45
|
|
|||||
09-841-140
![]() |
DWK Life Sciences
248390107 |
GL 32 |
Case of 10 for $295.87
|
|
|||||
Description
This threaded tube has a screw thread and can be incorporated into glass components by a further processing step.
- With DIN thread
Specifications
| Borosilicate Glass (3.3) | |
| Shoring in glass apparatuses | |
| Autoclavable |