Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
DWK Life Sciences™ DURAN™ YOUTILITY™ GL 45 Bottle Tag
Shop All DWK Life Sciences Products

Click to view available options
Color:
Black
Blue
Green
Orange
Purple
Red
White
Yellow
Description
These colorful GL 45 Bottle Tags offer an elegant solution for the marking of laboratory bottles.
- Available in different colors for marking of laboratory bottles and personalization
- Elastic Bottle Tags can be easily and securely attached around the neck, even with the screw cap in place
- Feature a graspable tab that makes them easy to remove with bare or gloved hands
- Chemically robust, heat resistant and totally reusable
- Fully compatible with all the GL 45 threads and therefore highly versatile

Specifications
Specifications
Quantity | 20 Tags/Pk. |
Type | Tag |
Color | Purple |
Material | Silicone |
For Use With (Application) | Color Coding and Identification |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction