Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ DUS1L Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA554783
Description
Immunogen sequence: LTVHGRTKEQ KGPLSGAASW EHIKAVRKAV AIPVFANGNI QCLQDVERCL RDTGVQGVMS AEGNLHNPAL FEGRSPAVWE LAEEYLDIVR EH Highest antigen sequence identity to the following orthologs: Mouse - 92%, Rat - 90%.
Catalyzes the synthesis of dihydrouridine, a modified base found in the D-loop of most tRNAs. Target protein has a region of similarity to tRNA dihydrouridine synthase.
Specifications
| DUS1L | |
| Polyclonal | |
| Unconjugated | |
| Dus1l | |
| 1110032N12Rik; dihydrouridine synthase 1 like; dihydrouridine synthase 1-like; dihydrouridine synthase 1-like (S. cerevisiae); DUS1; DUS1L; embryo-related protein; liver regeneration-related protein LRRG08/LRRG09; Pp3111; tRNA-dihydrouridine synthase 1-like; tRNA-dihydrouridine(16/17) synthase [NAD(P)(+)]-like; x85 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 64118 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Immunocytochemistry | |
| 0.3 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q6P1R4 | |
| Dus1l | |
| Recombinant protein corresponding to Human DUS1L. Recombinant protein control fragment (Product #RP-93410). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction