Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ DUS3L Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA559757
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA559757 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA559757 Supplier Invitrogen™ Supplier No. PA559757
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: TVEVDFVDIN VGCPIDLVYK KGGGCALMNR STKFQQIVRG MNQVLDVPLT VKIRTGVQER VNLAHRLLPE LRDWGVALVT LHGRSREQRY TKLADWQ Highest antigen sequence identity to the following orthologs: Mouse - 91%, Rat - 91%.

DUS3L gene ontology annotations related to this gene include flavin adenine dinucleotide binding; metal ion binding; tRNA dihydrouridine synthase activity.
TRUSTED_SUSTAINABILITY

Specifications

Antigen DUS3L
Applications Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
Classification Polyclonal
Concentration 0.3 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene DUS3L
Gene Accession No. Q3KRC5, Q91XI1, Q96G46
Gene Alias AI662135; AW557805; dihydrouridine synthase 3 like; dihydrouridine synthase 3-like; dihydrouridine synthase 3-like (S. cerevisiae); DUS3; DUS3L; putative zinc finger protein MA23; tRNA-dihydrouridine synthase 3-like; tRNA-dihydrouridine(47) synthase [NAD(P)(+)]-like
Gene Symbols DUS3L
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human DUS3L. Recombinant protein control fragment (Product #RP-99127).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 224907, 301122, 56931
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.