Learn More
DUS3L Rabbit anti-Human, Polyclonal, Bethyl Laboratories
Rabbit Polyclonal Antibody
Supplier: Bethyl Laboratories, Inc A304641AT

Description
The recommended shelf life for this product is 1 year from date of receipt.
DUS3L gene ontology annotations related to this gene include flavin adenine dinucleotide binding; metal ion binding; tRNA dihydrouridine synthase activity.Specifications
DUS3L | |
Polyclonal | |
Unconjugated | |
DUS3L | |
DUS3, tRNA-dihydrouridine(47) synthase [NAD(P)(+)]-like | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
56931 | |
4° C | |
Liquid |
Immunoprecipitation | |
1 mg/ml | |
phosphate, tris citrate with 0.09% sodium azide; pH 7-8 | |
Q96G46 | |
DUS3L | |
Between 175 and 225 | |
10 μL | |
Primary | |
Human | |
Antibody | |
IgG |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.