Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
DUSP10 Antibody, DyLight 405, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP130062V
Description
DUSP10 Polyclonal antibody specifically detects DUSP10 in Human samples. It is validated for Western BlotSpecifications
DUSP10 | |
Polyclonal | |
Western Blot | |
dual specificity phosphatase 10, dual specificity phosphatase MKP-5, EC 3.1.3.16, EC 3.1.3.48, MAP kinase phosphatase 5, Mitogen-activated protein kinase phosphatase 5, MKP-5dual specificity protein phosphatase 10, MKP5serine/threonine specific protein phosphatase | |
maps to a region between residue 432 and 482 of human dual specificity phosphatase 10 using the numbering given in entry NP_009138.1 | |
0.1 mL | |
Signal Transduction | |
11221 | |
Store at 4°C in the dark. | |
IgG |
Western Blot | |
DyLight 405 | |
50mM Sodium Borate | |
Rabbit | |
Immunogen affinity purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction