Learn More
DUSP10 Mouse anti-Human, Clone: OTI3D8, lyophilized, TrueMAB™

Mouse Monoclonal Antibody
Supplier: OriGene CF806754

Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
Dual specificity protein phosphatases inactivate their target kinases by dephosphorylating both the phosphoserine/threonine and phosphotyrosine residues. They negatively regulate members of the MAPK superfamily (MAPK/ERK, SAPK/JNK, p38), which is associated with cellular proliferation and differentiation. Different members of this family of dual specificity phosphatases show distinct substrate specificities for MAPKs, different tissue distribution and subcellular localization, and different modes of inducibility of their expression by extracellular stimuli. This gene product binds to and inactivates p38 and SAPK/JNK, but not MAPK/ERK. Its subcellular localization is unique; it is evenly distributed in both the cytoplasm and the nucleus. This gene is widely expressed in various tissues and organs, and its expression is elevated by stress stimuli. Three transcript variants encoding two different isoforms have been found for this gene.Specifications
DUSP10 | |
Monoclonal | |
Unconjugated | |
DUSP10 | |
MKP-5, MKP5 | |
Mouse | |
Affinity Chromatography | |
RUO | |
11221 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Lyophilized |
Western Blot | |
OTI3D8 | |
PBS with 8% trehalose and no preservative; pH 7.3 | |
Q9Y6W6 | |
DUSP10 | |
Human recombit protein fragment corresponding to amino acids 180-482 of human DUSP10 produced in E.coli. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG2a |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.