Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ DUSP10 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5109665
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5109665 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5109665 Supplier Invitrogen™ Supplier No. PA5109665
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: MPPSPLDDRV VVALSRPVRP QDLNLCLDSS YLGSANPGSN SHPPVIATTV VSLKAANLTY MPSSSGSARS LNCGCSSASC CTVATYDKDN QAQTQAIAAG TTTTAIGTST TCPANQMVNN NENTGSLSPS SGVGSPVSGT PKQLASIKII YPNDLAKKMT KCSKSHLPSQ GPVIIDCRPF MEYNKSHIQG AVHINCADKI SRRRLQQGKI TVLDLISCRE GKDSFKRIFS KEIIVYDENT NEPSRVMPSQ PLHIVLESLK REGKEPLVLK GGLSSFKQNH ENLCDNSLQL QECREVGGGA.

Microsomal long and very long chain fatty acid elongation uses malonyl-CoA as the 2-carbon donor and consists of 4 sequential reactions. GPSN2 catalyzes the final step, reducing trans-2, 3-enoyl-CoA to saturated acyl-CoA. Microsomal long and very long chain fatty acid elongation uses malonyl-CoA as the 2-carbon donor and consists of 4 sequential reactions. TER catalyzes the final step, reducing trans-2, 3-enoyl-CoA to saturated acyl-CoA.
TRUSTED_SUSTAINABILITY

Specifications

Antigen DUSP10
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 0.75 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene DUSP10
Gene Accession No. Q9ESS0, Q9Y6W6
Gene Alias 2610306G15Rik; AI158871; dual specificity phosphatase 10; dual specificity phosphatase MKP-5; dual specificity protein phosphatase 10; Dusp10; MAP kinase phosphatase 5; mitogen-activated protein kinase phosphatase 5; Mkp5; MKP-5; serine/threonine specific protein phosphatase
Gene Symbols DUSP10
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 1-300 of human DUSP10 (NP_009138.1).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 11221, 63953, 63995
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.