Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ DUSP10 Polyclonal Antibody
Goat Polyclonal Antibody
Supplier: Invitrogen™ PA131512
Description
Recommended positive controls: The peptide used to generate this antibody is available for purchase (GTX22272-PEP).. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
Microsomal long and very long chain fatty acid elongation uses malonyl-CoA as the 2-carbon donor and consists of 4 sequential reactions. GPSN2 catalyzes the final step, reducing trans-2, 3-enoyl-CoA to saturated acyl-CoA. Microsomal long and very long chain fatty acid elongation uses malonyl-CoA as the 2-carbon donor and consists of 4 sequential reactions. TER catalyzes the final step, reducing trans-2, 3-enoyl-CoA to saturated acyl-CoA.
Specifications
DUSP10 | |
Polyclonal | |
Unconjugated | |
DUSP10 | |
2610306G15Rik; AI158871; dual specificity phosphatase 10; dual specificity phosphatase MKP-5; dual specificity protein phosphatase 10; Dusp10; MAP kinase phosphatase 5; mitogen-activated protein kinase phosphatase 5; Mkp5; MKP-5; serine/threonine specific protein phosphatase | |
Goat | |
IgG fraction | |
RUO | |
11221 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Western Blot | |
0.5 mg/mL | |
TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3 | |
Q9Y6W6 | |
DUSP10 | |
Peptide with sequence RILTPKLMGVETVV, from C Terminus of the protein sequence according to NP_009138; NP_653329; NP_653330. | |
50 μg | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction