Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ DUSP10 Polyclonal Antibody

Goat Polyclonal Antibody
Supplier: Invitrogen™ PA518008
Description
This antibody is predicted to react with bovine, canine, mouse and rat based on sequence homology. This antibody is tested in Peptide ELISA: antibody detection limit dilution 8,000.
Microsomal long and very long chain fatty acid elongation uses malonyl-CoA as the 2-carbon donor and consists of 4 sequential reactions. GPSN2 catalyzes the final step, reducing trans-2, 3-enoyl-CoA to saturated acyl-CoA. Microsomal long and very long chain fatty acid elongation uses malonyl-CoA as the 2-carbon donor and consists of 4 sequential reactions. TER catalyzes the final step, reducing trans-2, 3-enoyl-CoA to saturated acyl-CoA.
Specifications
| DUSP10 | |
| Polyclonal | |
| Unconjugated | |
| DUSP10 | |
| 2610306G15Rik; AI158871; dual specificity phosphatase 10; dual specificity phosphatase MKP-5; dual specificity protein phosphatase 10; Dusp10; MAP kinase phosphatase 5; mitogen-activated protein kinase phosphatase 5; Mkp5; MKP-5; serine/threonine specific protein phosphatase | |
| Goat | |
| Ammonium sulfate precipitation | |
| RUO | |
| 11221 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3 | |
| Q9Y6W6 | |
| DUSP10 | |
| Synthetic peptide sequence (CRILTPKLMGVETVV) corresponding to the C-terminus amino acids of DUSP10 (aa 473-482). | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction