Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
DUSP11 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP258109
Description
DUSP11 Polyclonal specifically detects DUSP11 in Human samples. It is validated for Immunocytochemistry/Immunofluorescence.Specifications
DUSP11 | |
Polyclonal | |
Immunocytochemistry/Immunofluorescence 1-4 ug/ml | |
dual specificity phosphatase 11 (RNA/RNP complex 1-interacting), Dual specificity protein phosphatase 11, EC 3.1.3.-, Phosphatase that interacts with RNA/RNP complex 1, PIR1RNA/RNP complex-interacting phosphatase, RNA/RNP complex-1-interacting phosphatase, serine/threonine specific protein phosphatase | |
Rabbit | |
Affinity Purified | |
RUO | |
8446 | |
Human | |
IgG |
Immunocytochemistry, Immunofluorescence | |
Unconjugated | |
PBS, pH 7.2, containing 40% glycerol with 0.02% Sodium Azide | |
DUSP11 | |
This antibody was developed against a recombinant protein corresponding to the following amino acid sequence:MSQWHHPRSGWGRRRDFSGRSSAKKKGGNHIPERWKDYLPVGQRMPGTRFIAFKVPLQKSFEKKLAPEECFSPLDLFNKIREQNEELGLIIDLTYTQRYYKPEDLPETVPYLKIFTVGHQVPDDETIFKFKHAVNGFLKENKDNDKLIGV | |
100 μL | |
Primary | |
Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction