Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ DUSP12 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA530438
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA530438 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA530438 Supplier Invitrogen™ Supplier No. PA530438
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Recommended positive controls: A549, HeLa. Predicted reactivity: Mouse (87%), Rat (88%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

Dual specificity phosphatase (DUSP) inactivate their target kinases by dephosphorylating both the phosphoserine/threonine and phosphotyrosine residues. DUSPs can be divided into six subgroups on the basis of sequence similarity (PRLs, Cdc14 phosphatases, PTENs, myotubularins, MKPs, atypical DUSPs). DUSP inhibitors might be used to manipulate MAPK and cellular responses in both positive and negative ways. The regulated expression and activity of DUSP family members in different cells and tissues controls MAPK intensity and duration to determine the type of physiological response. DUSP12 (YVH1, GKAP) contains the consensus DUSP catalytic domain as well as an extended C-terminal domain of unknown function, thought to be related to its potential role in glucokinase regulation. It is localized in the cytoplasm and nucleus.
TRUSTED_SUSTAINABILITY

Specifications

Antigen DUSP12
Applications Western Blot
Classification Polyclonal
Concentration 0.94 mg/mL
Conjugate Unconjugated
Formulation 0.1M tris glycine with 20% glycerol and 0.01% thimerosal; pH 7
Gene Dusp12
Gene Accession No. Q9UNI6
Gene Alias 1190004O14Rik; AA027408; AW049275; dual specificity phosphatase 12; dual specificity phosphatase T-DSP4; Dual specificity phosphatase VH1; dual specificity protein phosphatase 12; Dual specificity tyrosine phosphatase YVH1; DUSP1; Dusp12; ESTM36; LMW-DSP4; mVH1; serine/threonine specific protein phosphatase; T-DSP4; VH1; YVH1; YVH1 protein-tyrosine phosphatase ortholog
Gene Symbols Dusp12
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the center region of human DUSP12. The exact sequence is proprietary.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 11266
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.