Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ DUSP12 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA589113
Description
Immunogen sequence: LMKTDQLPFE KAYEKLQILK PEAKMNEGFE WQLKLYQAMG YEVDTSSAIY KQYRLQKVTE KYPELQNLPQ ELFAVDPTTV SQGLKDEVLY KCRKCRRSLF RSSSILDHRE GSGPIAFAHK RMTPSSMLTT GRQAQCTSYF I; Positive Samples: Jurkat, A-549; Cellular Location: Nucleus.
Dual specificity phosphatase (DUSP) inactivate their target kinases by dephosphorylating both the phosphoserine/threonine and phosphotyrosine residues. DUSPs can be divided into six subgroups on the basis of sequence similarity (PRLs, Cdc14 phosphatases, PTENs, myotubularins, MKPs, atypical DUSPs). DUSP inhibitors might be used to manipulate MAPK and cellular responses in both positive and negative ways. The regulated expression and activity of DUSP family members in different cells and tissues controls MAPK intensity and duration to determine the type of physiological response. DUSP12 (YVH1, GKAP) contains the consensus DUSP catalytic domain as well as an extended C-terminal domain of unknown function, thought to be related to its potential role in glucokinase regulation. It is localized in the cytoplasm and nucleus.
Specifications
DUSP12 | |
Polyclonal | |
Unconjugated | |
Dusp12 | |
1190004O14Rik; AA027408; AW049275; dual specificity phosphatase 12; dual specificity phosphatase T-DSP4; Dual specificity phosphatase VH1; dual specificity protein phosphatase 12; Dual specificity tyrosine phosphatase YVH1; DUSP1; Dusp12; ESTM36; LMW-DSP4; mVH1; serine/threonine specific protein phosphatase; T-DSP4; VH1; YVH1; YVH1 protein-tyrosine phosphatase ortholog | |
Rabbit | |
Affinity chromatography | |
RUO | |
11266, 80915 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
ELISA, Western Blot | |
2.54 mg/mL | |
PBS with 50% glycerol and 0.01% thimerosal; pH 7.3 | |
Q9D0T2, Q9UNI6 | |
Dusp12 | |
Recombinant fusion protein containing a sequence corresponding to amino acids 130-270 of human DUSP12 (NP_0091711). | |
100 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction