Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
DUSP13 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$181.90 - $402.00
Specifications
Antigen | DUSP13 |
---|---|
Concentration | 0.5 mg/mL |
Dilution | Western Blot 0.5-2 ug/ml |
Applications | Western Blot |
Classification | Polyclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NB10056491T
![]() |
Novus Biologicals
NB10056491SS |
0.025 mg |
Each for $181.90
|
|
|||||
NB10056491
![]() |
Novus Biologicals
NB10056491 |
0.1 mg |
Each for $402.00
|
|
|||||
Description
DUSP13 Polyclonal specifically detects DUSP13 in Human, Rat, Bovine, Canine, Primate samples. It is validated for Western Blot.Specifications
DUSP13 | |
Western Blot 0.5-2 ug/ml | |
Polyclonal | |
Purified | |
RUO | |
Human, Rat, Bovine, Canine, Primate | |
NP_057448 | |
51207 | |
A portion of amino acids 150-200 of human DUSP13 was used as the immunogen. | |
Primary | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
0.5 mg/mL | |
Western Blot | |
Unconjugated | |
Rabbit | |
Cell Cycle and Replication | |
PBS containing 0.2% gelatin with 0.05% Sodium Azide | |
Branching-enzyme interacting DSP, dual specificity phosphatase 13, Dual specificity phosphatase SKRP4, dual specificity protein phosphatase 13, dual specificity protein phosphatase 13 isoform MDSP, dual-specificity phosphatase SKRP4, DUSP13A, DUSP13B, EC 3.1.3.16, EC 3.1.3.48, FLJ32450, MDSPbranching-enzyme interacting dual-specificity protein phosphatase, Muscle-restricted DSP, Testis- and skeletal-muscle-specific DSP, TMDPBEDPSKRP4 | |
DUSP13 | |
IgG | |
Protein G purified | |
The amino acid sequence used as immunogen is 100% conserved in human isoforms 1, 2 and 3. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title