Learn More
Invitrogen™ DUSP13 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA592995
Description
Immunogen sequence: MDSLQKQDLR RPKIHGAVQA SPYQPPTLAS LQRLLWVRQA ATLNHIDEVW PSLFLGDAYA ARDKSKLIQL GITHVVNAAA GKFQVDTGAK FYRGMSLEYY GIEADDNPFF DLSVYFLPVA RYIRAALSVP QGRVLVHCAM GVSRSATLVL AFLMICENMT LVEAIQTVQA HRNICPNSGF LRQLQVLDNR LGRETGRF; Positive Samples: Mouse skeletal muscle, Rat skeletal muscle; Cellular Location: Cytoplasm.
DUSP13, a novel DSP, is a member of highly conserved Protein-tyrosine phosphatase super family. Members of this family cooperate with protein kinases and play a vital role in cell cycle regulation. DUSP13 consists of a conserved catalytic domain and is expressed specifically in the testis and skeletal muscle. Based on mRNA localization and expression studies during development, DUSP13 is suggested to be expressed as 3 isoforms due to alternative splicing. Reports suggest DUSP13 might be involved in the regulation of meiosis and differentiation of testicular germ cells during spermatogenesis.
Specifications
DUSP13 | |
Polyclonal | |
Unconjugated | |
DUSP13 | |
BEDP; Branching-enzyme interacting DSP; branching-enzyme interacting dual-specificity protein phosphatase; Dsp13-like protein; dual specificity phosphatase 13; Dual specificity phosphatase SKRP4; dual specificity protein phosphatase 13; Dual specificity protein phosphatase 13 isoform A; Dual specificity protein phosphatase 13 isoform B; dual specificity protein phosphatase isoform MDSP; dual specificity tyrosine phosphatase TS-DSP6; dual-specificity tyrosine phosphatase TS-DSP6; DUSP13; Dusp13a; DUSP13B; FLJ32450; Gm1203; LMW-DSP6; Mdsp; muscle-restricted DSP; muscle-restricted dual specificity phosphatase; muscle-restricted dual specificity phosphatase; testis and skeletal muscle-specific dual specificity phosphatase; RP11-487I5.6; SKRP4; testis- and skeletal muscle-specific DSP; testis and skeletal muscle-specific dual specificity phosphatase; Testis- and skeletal-muscle-specific DSP; TMDP; TS-DSP6 | |
Rabbit | |
Affinity Chromatography | |
RUO | |
27389, 361002 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
ELISA, Western Blot | |
1.19 mg/mL | |
PBS with 50% glycerol and 0.01% thimerosal; pH 7.3 | |
Q6B8I0 | |
DUSP13 | |
Recombinant fusion protein containing a sequence corresponding to amino acids 1-198 of human DUSP13 (NP_0574483). | |
100 μL | |
Primary | |
Mouse, Rat | |
Antibody | |
IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.