Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DUSP13 Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501988022
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-8022 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-8022 Supplier Bioss Supplier No. BS7797R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

DUSP13, a novel DSP, is a member of highly conserved Protein-tyrosine phosphatase super family. Members of this family cooperate with protein kinases and play a vital role in cell cycle regulation. DUSP13 consists of a conserved catalytic domain and is expressed specifically in the testis and skeletal muscle. Based on mRNA localization and expression studies during development, DUSP13 is suggested to be expressed as 3 isoforms due to alternative splicing. Reports suggest DUSP13 might be involved in the regulation of meiosis and differentiation of testicular germ cells during spermatogenesis.

Specifications

Antigen DUSP13
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene DUSP13
Gene Accession No. Q6B8I1
Gene Alias BEDP; BOS_24228; Branching-enzyme interacting DSP; branching-enzyme interacting dual-specificity protein phosphatase; Dsp13-like protein; dual specificity phosphatase 13; Dual specificity phosphatase SKRP4; dual specificity protein phosphatase 13; Dual specificity protein phosphatase 13 isoform A; Dual specificity protein phosphatase 13 isoform B; dual specificity protein phosphatase isoform MDSP; Dual specificity tyrosine phosphatase TS-DSP6; dual-specificity tyrosine phosphatase TS-DSP6; Dusp13; Dusp13a; DUSP13B; FLJ32450; Gm1203; LMW-DSP6; LOC100356222; MDSP; Muscle-restricted DSP; muscle-restricted dual specificity phosphatase; muscle-restricted dual specificity phosphatase; testis and skeletal muscle-specific dual specificity phosphatase; RP11-487I5.6; SKRP4; Testis- and skeletal muscle-specific DSP; testis and skeletal muscle-specific dual specificity phosphatase; Testis- and skeletal-muscle-specific DSP; Tmdp; TS-DSP6
Gene Symbols DUSP13
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human DUSP13.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 51207
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.