Learn More
Invitrogen™ DUSP14 Polyclonal Antibody

Description
Positive test controls include: Hela, SW480, BT-474, SKOV3, Mouse kidney, Mouse brain. Immunogen sequence: MSSRGHSTLP RTLMAPRMIS EGDIGGIAQI TSSLFLGRGS VASNRHLLQA RGITCIVNAT IEIPNFNWPQ FEYVKVPLAD MPHAPIGLYF DTVADKIHSV SRKHGATLVH CAAGVSRSAT LCIAYLMKFH NVCLLEAYNW VKARRPVIRP NVGFWRQLID YERQLFGKST VKMVQTPYGI VPDVYEKESR HLMPYWGI.
Specifications
Specifications
| Antigen | DUSP14 |
| Applications | ELISA, Western Blot, Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 1.11 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 |
| Gene | DUSP14 |
| Gene Accession No. | O95147, Q9JLY7 |
| Gene Alias | 1110014C10Rik; 2310042C07Rik; D11Ertd395e; dual specificity phosphatase 14; dual specificity protein phosphatase 14; DUSP14; MAP kinase phosphatase 6; MAP kinase phsophatase 6; mitogen-activated protein kinase phosphatase 6; Mkp-1 like protein tyrosine phosphatase; MKP-1-like protein tyrosine phosphatase; MKP6; MKP-6; MKP-L |
| Gene Symbols | DUSP14 |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.