Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ DUSP15 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA567433
Description
Immunogen sequence: ICLCFGEEDPG PTQHPKEQLI MADVQVQLRP GSSSCTLSAS TERPDGSSTP GNPDGITHLQ CSCLHPKRA Highest antigen sequence identity to the following orthologs - mouse 31%, rat 35%.
The protein encoded by this gene belongs to the non-receptor class of the protein-tyrosine phosphatase family. The encoded protein has both protein-tyrosine phophatase activity and serine/threonine-specific phosphatase activity, and therefore is known as a dual specificity phosphatase. Three transcript variants encoding two different isoforms have been found for this gene.
Specifications
DUSP15 | |
Polyclonal | |
Unconjugated | |
Dusp15 | |
AI851682; C20orf57; dual specificity phosphatase 15; dual specificity phosphatase-like 15; dual specificity protein phosphatase 15; Dual specificity protein phosphatase T-DSP10; DUSP15; FLJ40111; LMW-DSP10; LOW QUALITY PROTEIN: dual specificity protein phosphatase 15; RP11-243J16.11; T-DSP10; Vaccinia virus VH1-related dual-specific protein phosphatase Y; VH1-related member Y; VHY | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
128853 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Immunocytochemistry | |
0.3 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q9H1R2 | |
Dusp15 | |
Recombinant Human DUSP15. Recombinant protein control fragment (Product #RP-108094). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction