Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ DUSP16 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA587683
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA587683 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA587683 Supplier Invitrogen™ Supplier No. PA587683
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: SLHGFSSSED ALEYYKPSTT LDGTNKLCQF SPVQELSEQT PETSPDKEEA SIPKKLQTAR PSDSQSKRLH SVRTSSSGTA QRSLLSPLHR SGSVEDNYHT SFLFGLSTSQ QHLTKSAGLG LKGWHSDILA PQTSTPSLTS SWYFATESSH FYSASAIYGG SASYSAYSCS QLPTCGDQVY SVRRRQKPSD RADSRRSWHE ESPFEKQFKR RSCQMEFGES IMSENRSREE LGKVGSQSSF SGSMEIIEVS; Positive Samples: HeLa, A-549, K-562, HepG2, Mouse liver; Cellular Location: Cytoplasm, Cytoplasmic vesicle, Nucleus.

Dual-specificity phosphatases constitute a large heterogeneous subgroup of the type I cysteine-based protein-tyrosine phosphatase superfamily. DUSPs are characterized by their ability to dephosphorylate both tyrosine and serine/threonine residues. DUSP16 belongs to a class of DUSPs, designated MKPs, that dephosphorylate MAPK proteins ERK, JNK, and p38 with specificity distinct from that of individual MKP proteins. MKPs contain a highly conserved C-terminal catalytic domain and an N-terminal Cdc25-like domain. MAPK activation cascades mediate various physiologic processes, including cellular proliferation, apoptosis, differentiation, and stress responses.
TRUSTED_SUSTAINABILITY

Specifications

Antigen DUSP16
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 0.33 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene DUSP16
Gene Accession No. Q9BY84
Gene Alias 3830417M17Rik; AW558566; D6Ertd213e; dual specificity phosphatase 16; dual specificity protein phosphatase 16; Dusp16; KIAA1700; MAP kinase phosphatase 7; MAP kinase phosphatase-7; map kinase phosphatase-M; MAPK phosphatase-7; MGC129701; MGC129702; mitogen-activated protein kinase phosphatase 7; Mkp7; MKP-7; Mkpm
Gene Symbols DUSP16
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 416-665 of human DUSP16 (NP_0851431).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 297682, 70686, 80824
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.