Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ DUSP16 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA599008
Description
DUSP16 Polyclonal Antibody for Western Blot, IHC (P), ELISA
Dual-specificity phosphatases constitute a large heterogeneous subgroup of the type I cysteine-based protein-tyrosine phosphatase superfamily. DUSPs are characterized by their ability to dephosphorylate both tyrosine and serine/threonine residues. DUSP16 belongs to a class of DUSPs, designated MKPs, that dephosphorylate MAPK proteins ERK, JNK, and p38 with specificity distinct from that of individual MKP proteins. MKPs contain a highly conserved C-terminal catalytic domain and an N-terminal Cdc25-like domain. MAPK activation cascades mediate various physiologic processes, including cellular proliferation, apoptosis, differentiation, and stress responses.
Specifications
DUSP16 | |
Polyclonal | |
Unconjugated | |
DUSP16 | |
3830417M17Rik; AW558566; D6Ertd213e; dual specificity phosphatase 16; dual specificity protein phosphatase 16; Dusp16; KIAA1700; MAP kinase phosphatase 7; MAP kinase phosphatase-7; map kinase phosphatase-M; MAPK phosphatase-7; MGC129701; MGC129702; mitogen-activated protein kinase phosphatase 7; Mkp7; MKP-7; Mkpm | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
80824 | |
-20°C or -80°C if preferred | |
Liquid |
ELISA, Immunohistochemistry (Paraffin), Western Blot | |
0.52 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q9BY84 | |
DUSP16 | |
Recombinant Human Dual specificity protein phosphatase 16 protein (416-665AA). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction