Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
DUSP18 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$377.03 - $728.30
Specifications
| Antigen | DUSP18 |
|---|---|
| Applications | Immunocytochemistry, Immunofluorescence |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Host Species | Rabbit |
Description
DUSP18 Polyclonal specifically detects DUSP18 in Human samples. It is validated for Immunocytochemistry/Immunofluorescence.Specifications
| DUSP18 | |
| Polyclonal | |
| Rabbit | |
| Protein Phosphatase | |
| PBS, pH 7.2, containing 40% glycerol with 0.02% Sodium Azide | |
| 150290 | |
| This antibody was developed against a recombinant protein corresponding to the following amino acid sequence:APSCAFPVQFRQPSVSGLSQITKSLYISNGVAATN | |
| Primary | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
| Immunocytochemistry, Immunofluorescence | |
| Unconjugated | |
| RUO | |
| Human | |
| bK963H5.1, DSP18, dual specificity phosphatase 18, dual specificity protein phosphatase 18, DUSP20, EC 3.1.3.16, EC 3.1.3.48, LMWDSP20, LMW-DSP20, Low molecular weight dual specificity phosphatase 20, MGC32658 | |
| DUSP18 | |
| IgG | |
| Affinity Purified |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title