Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ DUSP18 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA565657
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA565657 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA565657 Supplier Invitrogen™ Supplier No. PA565657
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: APSCAFPVQFR QPSVSGLSQI TKSLYISNGV AATN Highest antigen sequence identity to the following orthologs - mouse 74%, rat 71%.

DUSP18 (E.C 3.1.3.16), a novel DSP, is a member of highly conserved protein-tyrosine phosphatase super family. Members of this family cooperate with protein kinases and play a vital role in cell cycle regulation and mitogenic signal transduction. DUSP18 is a low molecular weight protein consisting of conserved N-terminal DSP domain but lacks CH2 domain. Studies reveal that DUSP18 is a unique DSP that interacts and dephosphorylates SAPK/JNK. Reports suggest it inhibits SAPK/JNK invivo and might serve an important role in the regulation of SAPK/JNK pathway. Northern Blot analysis detected a ubiquitous expression of DUSP18.
TRUSTED_SUSTAINABILITY

Specifications

Antigen DUSP18
Applications Immunocytochemistry
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene DUSP18
Gene Accession No. Q8NEJ0
Gene Alias bK963H5.1; DSP18; dual specificity phosphatase 18; dual specificity protein phosphatase 18; DUSP18; DUSP20; hypothetical protein LOC505912; LMWDSP20; LMW-DSP20; Low molecular weight dual specificity phosphatase 20
Gene Symbols DUSP18
Host Species Rabbit
Immunogen Recombinant Human DUSP18. Recombinant protein control fragment (Product #RP-106334).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 150290
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.