Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ DUSP18 Polyclonal Antibody

Catalog No. PIPA523128
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA523128 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA523128 Supplier Invitrogen™ Supplier No. PA523128

Rabbit Polyclonal Antibody

Suggested positive control: human testis protein, human or rat testis lysate, rat testis protein.

DUSP18 (E.C 3.1.3.16), a novel DSP, is a member of highly conserved protein-tyrosine phosphatase super family. Members of this family cooperate with protein kinases and play a vital role in cell cycle regulation and mitogenic signal transduction. DUSP18 is a low molecular weight protein consisting of conserved N-terminal DSP domain but lacks CH2 domain. Studies reveal that DUSP18 is a unique DSP that interacts and dephosphorylates SAPK/JNK. Reports suggest it inhibits SAPK/JNK invivo and might serve an important role in the regulation of SAPK/JNK pathway. Northern Blot analysis detected a ubiquitous expression of DUSP18.
TRUSTED_SUSTAINABILITY

Specifications

Antigen DUSP18
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.2% gelatin and 0.05% sodium azide
Gene DUSP18
Gene Accession No. Q5BIP9, Q8NEJ0
Gene Alias bK963H5.1; DSP18; dual specificity phosphatase 18; dual specificity protein phosphatase 18; DUSP18; DUSP20; hypothetical protein LOC505912; LMWDSP20; LMW-DSP20; Low molecular weight dual specificity phosphatase 20
Gene Symbols DUSP18
Host Species Rabbit
Immunogen Synthetic peptide (9 PVQFRQPSVSGLSQ 22) of DUSP18 protein.
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 150290, 505912
Target Species Human, Monkey, Bovine
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.