Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
DUSP19 Polyclonal Antibody
DUSP19 Polyclonal Antibody
Supplier: Thermo Scientific PA554428
Description
DUSP19 Polyclonal Antibody for Western Blot, IHC

Specifications
DUSP19 | |
Polyclonal | |
Unconjugated | |
DUSP19 | |
5930436K22Rik; C79103; dual specificity phosphatase 19; dual specificity phosphatase TS-DSP1; Dual specificity protein phosphatase 19; DUSP17; DUSP19; LMWDSP3; LMW-DSP3; low molecular weight dual specificity phosphatase 3; Protein phosphatase SKRP1; SAPK pathway-regulating phosphatase 1; SKRP1; Stress-activated protein kinase pathway-regulating phosphatase 1; TS-DSP1 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
142679 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.2 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q8WTR2 | |
DUSP19 | |
Recombinant protein corresponding to Human DUSP19. Recombinant protein control fragment (Product #RP-91570). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction