Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ DUSP21 Polyclonal Antibody
GREENER_CHOICE

Catalog No. FEPA5144184
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FEPA5144184 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. FEPA5144184 Supplier Invitrogen™ Supplier No. PA5144184
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

DUSP21 Polyclonal Antibody for IHC (P), ELISA

This gene encodes a member of the dual specificity phosphatase family, specifically the low molecular weight dual specificity phosphatase family. The encoded protein localizes to both the cytoplasm and the nucleus and functions to remove phosphate groups from phosphotyrosine and phosphothreonine residues.
TRUSTED_SUSTAINABILITY

Specifications

Antigen DUSP21
Applications ELISA, Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 3 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.03% Proclin 300; pH 7.4
Gene DUSP21
Gene Accession No. Q9H596
Gene Alias BJ-HCC-26 tumor antigen; dual specificity phosphatase 21; dual specificity protein phosphatase 21; DUSP21; LMWDSP21; LMW-DSP21; Low molecular weight dual specificity phosphatase 21
Gene Symbols DUSP21
Host Species Rabbit
Immunogen Recombinant Human Dual specificity protein phosphatase 21 protein (1-190AA).
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 63904
Target Species Human
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.