Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
DUSP22 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP216239
Description
DUSP22 Polyclonal specifically detects DUSP22 in Human samples. It is validated for Western Blot.Specifications
DUSP22 | |
Polyclonal | |
Unconjugated | |
PBS (pH 7.0), 1.0% BSA and 20% Glycerol with 0.01% Thimerosal | |
dual specificity phosphatase 22, dual specificity protein phosphatase 22, EC 3.1.3.16, EC 3.1.3.48, FLJ35864, homolog of mouse dual specificity phosphatase LMW-DSP2, JKAP, JNK-stimulating phosphatase 1, JSP-1, JSP1JNK-stimulatory phosphatase-1, LMWDSP2, LMW-DSP2, Low molecular weight dual specificity phosphatase 2, Mitogen-activated protein kinase phosphatase x, MKP-x, MKPXMAP kinase phosphatase x, VHX | |
Rabbit | |
21 kDa | |
0.1 mL | |
GPCR, MAP Kinase Signaling, Protein Phosphatase | |
56940 | |
Human | |
IgG |
Western Blot | |
0.83mg/mL | |
Western Blot 1:500-1:3000 | |
Q9NRW4 | |
DUSP22 | |
Recombinant protein encompassing a sequence within the center region of human DUSP22. The exact sequence is proprietary. | |
Affinity Purified | |
RUO | |
Primary | |
Human. Expected to cross react based on sequence identity: Mouse (93%), Rat (91%). | |
Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction