Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ DUSP22 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA533487
Description
Percent identity with other species by BLAST analysis: Human, Gorilla, Gibbon, Monkey, Marmoset, Bovine, Horse, Rabbit (100%) Mouse, Rat (94%) Elephant, Chicken (88%) Xenopus (82%).
Mitogen-activated protein (MAP) kinases are a large class of proteins involved in signal transduction pathways that are activated by a range of stimuli and mediate a number of physiological and pathological changes in the cell. Dual specificity phosphatases (DSPs) are a subclass of the protein tyrosine phosphatase (PTP) gene superfamily, which are selective for dephosphorylating critical phosphothreonine and phosphotyrosine residues within MAP kinases. DSP gene expression is induced by a host of growth factors and/or cellular stresses, thereby negatively regulating MAP kinase superfamily members including MAPK/ERK, SAPK/JNK and p38. DUSP22 dephosphorylates ERK2 MAP kinase and JNK. DUSP22 displays highest in thymus, but it is also detectable in monocytes and lymphocytes.
Specifications
| DUSP22 | |
| Polyclonal | |
| Unconjugated | |
| Dusp22 | |
| DKFZp586F2224; dual specificity phosphatase 22; dual specificity protein phosphatase 22; Dusp22; homolog of mouse dual specificity phosphatase LMW-DSP2; JKAP; JNK-stimulating phosphatase 1; JNK-stimulatory phosphatase-1; JSP 1; JSP1; JSP-1; LMWDSP2; LMW-DSP2; low molecular weight dual specificity phosphatase 2; MAP kinase phosphatase x; mi; mitogen-activated protein kinase phosphatase x; MKPX; MKP-x; PYST2; VHX | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 100054792, 100353877, 56940, 787471 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 1 mg/mL | |
| PBS with 0.1% sodium azide | |
| Q9NRW4 | |
| Dusp22 | |
| Synthetic 17 amino acid peptide from near N-terminus of human DUSP22. | |
| 50 μg | |
| Primary | |
| Human, Rabbit, Monkey, Bovine, Horse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction