Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ DUSP22 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA533489
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA533489 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA533489 Supplier Invitrogen™ Supplier No. PA533489
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Percent identity with other species by BLAST analysis: Human, Gorilla, Gibbon, Monkey, Marmoset, Elephant, Bovine, Horse, Rabbit (100%) Mouse, Rat, Panda, Turkey, Chicken (94%).

Mitogen-activated protein (MAP) kinases are a large class of proteins involved in signal transduction pathways that are activated by a range of stimuli and mediate a number of physiological and pathological changes in the cell. Dual specificity phosphatases (DSPs) are a subclass of the protein tyrosine phosphatase (PTP) gene superfamily, which are selective for dephosphorylating critical phosphothreonine and phosphotyrosine residues within MAP kinases. DSP gene expression is induced by a host of growth factors and/or cellular stresses, thereby negatively regulating MAP kinase superfamily members including MAPK/ERK, SAPK/JNK and p38. DUSP22 dephosphorylates ERK2 MAP kinase and JNK. DUSP22 displays highest in thymus, but it is also detectable in monocytes and lymphocytes.
TRUSTED_SUSTAINABILITY

Specifications

Antigen DUSP22
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.1% sodium azide
Gene Dusp22
Gene Accession No. Q9NRW4
Gene Alias DKFZp586F2224; dual specificity phosphatase 22; dual specificity protein phosphatase 22; Dusp22; homolog of mouse dual specificity phosphatase LMW-DSP2; JKAP; JNK-stimulating phosphatase 1; JNK-stimulatory phosphatase-1; JSP 1; JSP1; JSP-1; LMWDSP2; LMW-DSP2; low molecular weight dual specificity phosphatase 2; MAP kinase phosphatase x; mi; mitogen-activated protein kinase phosphatase x; MKPX; MKP-x; PYST2; VHX
Gene Symbols Dusp22
Host Species Rabbit
Immunogen Synthetic 17 amino acid peptide from internal region of human DUSP22.
Purification Method Antigen affinity chromatography
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 100054792, 100353877, 56940, 787471
Target Species Human, Rabbit, Monkey, Bovine, Horse
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.