Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
DUSP23 Antibody (OTI3C10), DyLight 550, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP271976R
Description
DUSP23 Monoclonal specifically detects DUSP23 in Human, Mouse, Rat samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.Specifications
DUSP23 | |
Monoclonal | |
DyLight 550 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
DUSP23 | |
Full length recombinant protein of human DUSP23 (NP_060293) produced in HEK293T cells. | |
0.1 mL | |
Primary | |
Human, Mouse, Rat | |
IgG2a |
Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence | |
OTI3C10 | |
Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence | |
dual specificity phosphatase 23, dual specificity protein phosphatase 23, DUSP25, EC 3.1.3.16, EC 3.1.3.48, FLJ20442, LDP3, LDP-3, Low molecular mass dual specificity phosphatase 3, low-molecular-mass dual-specificity phosphatase 3, MOSP, RP11-190A12.1, VH1-like member Z, VH1-like phosphatase Z, VHZ | |
Mouse | |
Affinity Purified | |
GPCR, Protein Phosphatase | |
54935.0 | |
Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction